Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025108.1 | NP_001020279.1 | 15 | 2716 | ||
NM_002285.2 | NP_002276.2 | 15 | 2733 | |||
XM_005263943.3 | XP_005264000.2 | 14 | 2610 | |||
XM_005263945.3 | XP_005264002.1 | 6 | 1967 | |||
XM_011511169.2 | XP_011509471.1 | 15 | 2713 | |||
XM_011511170.2 | XP_011509472.1 | 16 | 2852 | |||
XM_011511171.2 | XP_011509473.1 | 15 | 2716 | |||
XM_011511172.2 | XP_011509474.1 | 14 | 2668 | |||
XM_011511173.2 | XP_011509475.1 | 15 | 2729 | |||
XM_011511174.2 | XP_011509476.1 | 15 | 2673 | |||
XM_011511175.2 | XP_011509477.1 | 17 | 2963 | |||
XM_011511176.2 | XP_011509478.1 | 14 | 2654 | |||
XM_011511177.2 | XP_011509479.1 | 15 | 2744 | |||
XM_011511178.2 | XP_011509480.1 | 5 | 1475 | |||
XM_011511179.2 | XP_011509481.1 | 5 | 1472 | |||
XM_017004085.1 | XP_016859574.1 | 6 | 1964 | |||
XM_017004086.1 | XP_016859575.1 | 6 | 2445 | |||
XM_017004087.1 | XP_016859576.1 | 6 | 2442 | |||
GenBank mRNA | AB209634.1 | - | 2 | 435 | ||
AK092327.1 | - | 16 | 2805 | |||
AK290468.1 | - | 14 | 2591 | |||
AK297407.1 | - | 16 | 2981 | |||
BC136579.1 | - | 13 | 2516 | 9020569 | ||
BC144266.1 | - | 14 | 2591 | 9052786 | ||
U34360.1 | - | 13 | 2551 |