Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025108.1 | NP_001020279.1 | 17 | 3121 | ||
NM_002285.2 | NP_002276.2 | 17 | 3138 | |||
XM_005263943.3 | XP_005264000.2 | 16 | 3015 | |||
XM_005263945.3 | XP_005264002.1 | 8 | 2372 | |||
XM_011511169.2 | XP_011509471.1 | 17 | 3118 | |||
XM_011511170.2 | XP_011509472.1 | 18 | 3257 | |||
XM_011511171.2 | XP_011509473.1 | 17 | 3121 | |||
XM_011511172.2 | XP_011509474.1 | 16 | 3073 | |||
XM_011511173.2 | XP_011509475.1 | 17 | 3134 | |||
XM_011511174.2 | XP_011509476.1 | 17 | 3078 | |||
XM_011511175.2 | XP_011509477.1 | 19 | 3368 | |||
XM_011511176.2 | XP_011509478.1 | 16 | 3059 | |||
XM_011511177.2 | XP_011509479.1 | 17 | 3149 | |||
XM_011511178.2 | XP_011509480.1 | 7 | 1880 | |||
XM_011511179.2 | XP_011509481.1 | 7 | 1877 | |||
XM_017004085.1 | XP_016859574.1 | 8 | 2369 | |||
XM_017004086.1 | XP_016859575.1 | 8 | 2850 | |||
XM_017004087.1 | XP_016859576.1 | 8 | 2847 | |||
GenBank mRNA | AB209634.1 | - | 4 | 840 | ||
BC136579.1 | - | 15 | 2921 | 9020569 | ||
BC144266.1 | - | 16 | 2996 | 9052786 | ||
U34360.1 | - | 15 | 2956 |