Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261456.1 | NP_001248385.1 | 10 | 2207 | ||
NM_001261457.1 | NP_001248386.1 | 9 | 1979 | |||
NM_002348.3 | NP_002339.2 | 10 | 2249 | |||
XM_011509548.1 | XP_011507850.1 | 11 | 2312 | |||
XM_011509549.1 | XP_011507851.1 | 11 | 2309 | |||
XM_011509550.1 | XP_011507852.1 | 11 | 2349 | |||
XM_011509552.1 | XP_011507854.1 | 10 | 2240 | |||
XM_011509556.1 | XP_011507858.1 | 10 | 2039 | |||
XM_011509560.1 | XP_011507862.1 | 10 | 1997 | |||
XM_017001299.1 | XP_016856788.1 | 11 | 2267 | |||
GenBank mRNA | AF244129.1 | - | 10 | 2162 | ||
AK292048.1 | - | 10 | 2149 | |||
AY007142.1 | - | 9 | 1569 | |||
BC027920.1 | - | 9 | 1815 | 5225850 | ||
BC062589.1 | - | 6 | 1191 | |||
BC121049.1 | - | 9 | 1914 | 40117946 | ||
BX090162.1 | - | 3 | 515 | |||
L42621.1 | - | 10 | 2000 |