Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318758.1 | NP_001305687.1 | 2 | 341 | ||
NM_002504.5 | NP_002495.2 | 2 | 341 | |||
NM_147134.3 | NP_667345.1 | 2 | 341 | |||
GenBank mRNA | AF332009.1 | - | 2 | 246 | ||
AK225750.1 | - | 2 | 214 | |||
AK291643.1 | - | 2 | 231 | |||
AL832342.1 | - | 2 | 223 | |||
BC012151.1 | - | 2 | 211 | 4558442 | ||
BP871782.1 | - | 2 | 283 | |||
DA093124.1 | - | 2 | 339 | |||
DA290058.1 | - | 2 | 287 | |||
DB076795.1 | - | 2 | 341 | |||
U15306.1 | - | 1 | 136 |