Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129889.2 | NP_001123361.1 | 2 | 843 | ||
NM_001322184.1 | NP_001309113.1 | 3 | 419 | |||
NM_001322185.1 | NP_001309114.1 | 2 | 365 | |||
NM_001322186.1 | NP_001309115.1 | 4 | 634 | |||
NM_001922.4 | NP_001913.2 | 2 | 843 | |||
XM_011521049.1 | XP_011519351.1 | 4 | 1467 | |||
XM_017020401.1 | XP_016875890.1 | 2 | 846 | |||
GenBank mRNA | AJ000503.1 | - | 2 | 815 | ||
AK293115.1 | - | 2 | 626 | |||
BC028311.1 | - | 2 | 690 | 4705935 | ||
D17547.1 | - | 2 | 830 | |||
DQ902581.1 | - | 2 | 708 | |||
L18967.1 | - | 2 | 800 | |||
S69231.1 | - | 2 | 765 |