Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHEX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002729.4 | NP_002720.1 | 2 | 494 | ||
GenBank mRNA | AK309463.1 | - | 2 | 1261 | ||
AK314891.1 | - | 2 | 463 | |||
BC014336.1 | - | 2 | 467 | 4046641 | ||
BC015110.1 | - | 2 | 467 | 4048691 | ||
BC050638.1 | - | 2 | 434 | 6174612 | ||
L16499.1 | - | 2 | 466 | |||
X67235.1 | - | 2 | 438 | |||
Z21533.1 | - | 1 | 409 |