Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134367.3 | NP_001127839.2 | 11 | 1601 | ||
NM_003043.5 | NP_003034.2 | 11 | 1605 | |||
NR_103507.2 | 10 | 1480 | ||||
XM_006713307.2 | XP_006713370.1 | 10 | 1409 | |||
XM_011534030.2 | XP_011532332.1 | 10 | 1362 | |||
XM_011534031.2 | XP_011532333.1 | 12 | 1733 | |||
GenBank mRNA | AB209172.1 | - | 10 | 1406 | ||
AK303649.1 | - | 10 | 1480 | |||
AK308417.1 | - | 9 | 1081 | |||
BC137128.1 | - | 11 | 1552 | 9021125 | ||
BC137129.1 | - | 11 | 1548 | 9021126 | ||
U09220.1 | - | 9 | 1280 | |||
U16120.1 | - | 12 | 1656 | |||
Z18956.1 | - | 9 | 1299 |