Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003106.3 | NP_003097.1 | 1 | 2184 | ||
GenBank mRNA | CN430685.1 | - | 1 | 127 |