Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 9 | 1140 | ||
NM_003151.3 | NP_003142.1 | 9 | 1191 | |||
XM_006712719.3 | XP_006712782.1 | 9 | 1186 | |||
XM_011511705.2 | XP_011510007.1 | 12 | 1337 | |||
XM_017004784.1 | XP_016860273.1 | 9 | 945 | |||
GenBank mRNA | AK299976.1 | - | 9 | 912 | ||
AK300868.1 | - | 9 | 944 | |||
AK301172.1 | - | 7 | 615 | |||
BC031212.1 | - | 9 | 1112 | 4830583 | ||
L78440.1 | - | 9 | 957 |