Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 15, 16 | 2073 | ||
NM_001178079.1 | NP_001171550.1 | 15, 16 | 1999 | |||
NM_001178080.1 | NP_001171551.1 | 14, 15 | 1917 | |||
NM_001178081.1 | NP_001171552.1 | 13, 14 | 1778 | |||
NM_003153.4 | NP_003144.3 | 15, 16 | 2054 | |||
NR_033659.1 | 14, 15 | 1915 | ||||
XM_006719575.2 | XP_006719638.1 | 16, 17 | 2051 | |||
XM_011538703.2 | XP_011537005.1 | 16, 17 | 2128 | |||
XM_011538704.2 | XP_011537006.1 | 17, 18 | 2139 | |||
XM_011538705.2 | XP_011537007.1 | 17, 18 | 2249 | |||
XM_011538706.2 | XP_011537008.1 | 17, 18 | 2105 | |||
XM_011538707.2 | XP_011537009.1 | 16, 17 | 2090 | |||
XM_011538708.2 | XP_011537010.1 | 15, 16 | 1932 | |||
XM_011538709.2 | XP_011537011.1 | 16, 17 | 1966 | |||
GenBank mRNA | AB103089.1 | - | 9, 10 | 1226 | ||
AK290431.1 | - | 15, 16 | 2002 | |||
AK291044.1 | - | 15, 16 | 2000 | |||
AK298721.1 | - | 13, 14 | 1598 | |||
AK299094.1 | - | 14, 15 | 1851 | |||
AK308751.1 | - | 15, 16 | 1894 | |||
AK312471.1 | - | 15, 16 | 1920 | |||
AK316142.1 | - | 14, 15 | 1861 | |||
AK316236.1 | - | 14, 15 | 1692 | |||
BC075852.1 | - | 15, 16 | 1855 | 5530399 | ||
BT007380.1 | - | 14, 15 | 1748 | |||
U16031.1 | - | 15, 16 | 1913 |