Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172085.1 | NP_001165556.1 | 7 | 1234 | ||
NM_003194.4 | NP_003185.1 | 8 | 1436 | |||
GenBank mRNA | AK298272.1 | - | 9 | 1318 | ||
AK304648.1 | - | 7 | 1221 | |||
BC109053.1 | - | 8 | 1347 | 40032492 | ||
BC109054.1 | - | 9 | 1201 | 40032493 | ||
BC110341.1 | - | 8 | 1396 | 5761426 | ||
M55654.1 | - | 8 | 1398 | |||
X54993.1 | - | 7 | 1243 |