Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003222.3 | NP_003213.1 | 7 | 2329 | ||
GenBank mRNA | BC035664.1 | - | 7 | 2289 | 5723548 | |
BC051829.1 | - | 7 | 2328 | 6148554 | ||
U85658.1 | - | 7 | 2252 |