Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001816.3 | NP_001807.2 | 6 | 2038 | ||
GenBank mRNA | BC026263.1 | - | 6 | 2002 | 4618311 | |
D90064.1 | - | 6 | 2023 | |||
M33326.1 | - | 6 | 2023 |