Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258379.1 | NP_001245308.1 | 25 | 4630 | ||
NM_001258380.1 | NP_001245309.1 | 24 | 4284 | |||
NM_001321103.1 | NP_001308032.1 | 26 | 5014 | |||
NM_001321104.1 | NP_001308033.1 | 26 | 4975 | |||
NM_001321105.1 | NP_001308034.1 | 25 | 4867 | |||
NM_001321106.1 | NP_001308035.1 | 25 | 4855 | |||
NM_001321107.1 | NP_001308036.1 | 24 | 4522 | |||
NM_001321108.1 | NP_001308037.1 | 25 | 4642 | |||
NM_003615.4 | NP_003606.3 | 25 | 4656 | |||
NR_135541.1 | 24 | 4680 | ||||
NR_135542.1 | 24 | 4455 | ||||
NR_135543.1 | 23 | 4347 | ||||
NR_135544.1 | 25 | 4788 | ||||
XM_005265598.4 | XP_005265655.1 | 25 | 4896 | |||
XM_011534258.1 | XP_011532560.1 | 25 | 4870 | |||
XM_011534261.2 | XP_011532563.1 | 25 | 5132 | |||
XM_011534265.1 | XP_011532567.1 | 24 | 4498 | |||
XM_017007528.1 | XP_016863017.1 | 19 | 3515 | |||
XM_017007529.1 | XP_016863018.1 | 19 | 3497 | |||
GenBank mRNA | AF047033.1 | - | 25 | 4657 | ||
AF053755.1 | - | 21 | 3962 | |||
AF089726.1 | - | 24 | 4285 | |||
CR627428.1 | - | 18 | 3346 |