Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001858.5 | NP_001849.2 | 17, 18 | 1466 | ||
XM_011535437.2 | XP_011533739.1 | 17, 18 | 1733 | |||
XM_011535438.2 | XP_011533740.1 | 17, 18 | 1461 | |||
XM_017010254.1 | XP_016865743.1 | 16, 17 | 1679 | |||
XM_017010255.1 | XP_016865744.1 | 17, 18 | 1672 | |||
XM_017010257.1 | XP_016865746.1 | 16, 17 | 1607 | |||
XM_017010258.1 | XP_016865747.1 | 16, 17 | 1340 | |||
XM_017010259.1 | XP_016865748.1 | 17, 18 | 1732 | |||
GenBank mRNA | BC113362.1 | - | 17, 18 | 1434 | 8322414 | |
BC113364.1 | - | 17, 18 | 1434 | 8322416 | ||
D38163.1 | - | 17, 18 | 1466 | |||
L12347.1 | - | 13, 14 | 1034 | |||
M63597.1 | - | 16, 17 | 1275 |