Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004739.3 | NP_004730.2 | 6, 7 | 887 | ||
XM_017018561.1 | XP_016874050.1 | 6, 7 | 739 | |||
GenBank mRNA | AB016591.1 | - | 6, 7 | 820 | ||
AF295807.1 | - | 6, 7 | 497 | |||
AK301569.1 | - | 6, 7 | 722 | |||
AL561056.3 | - | 6, 7 | 887 | |||
BC023656.2 | - | 6, 7 | 683 | |||
BC053650.1 | - | 6, 7 | 705 | 5531965 | ||
CR749481.1 | - | 4, 5 | 408 |