Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 5 | 695 | ||
NM_001258401.2 | NP_001245330.1 | 5 | 641 | |||
NM_001258402.1 | NP_001245331.1 | 5 | 695 | |||
NM_001258403.1 | NP_001245332.1 | 5 | 767 | |||
NM_001267615.1 | NP_001254544.1 | 5 | 641 | |||
XM_006719700.1 | XP_006719763.1 | 5 | 696 | |||
XM_011539010.1 | XP_011537312.1 | 5 | 696 | |||
XM_011539011.1 | XP_011537313.1 | 5 | 641 | |||
XM_017020240.1 | XP_016875729.1 | 5 | 723 | |||
XM_017020241.1 | XP_016875730.1 | 5 | 1055 | |||
XM_017020242.1 | XP_016875731.1 | 5 | 710 | |||
GenBank mRNA | AB018282.1 | - | 7 | 775 | ||
AF069512.1 | - | 5 | 532 | |||
AF107099.1 | - | 5 | 531 | |||
AK128321.1 | - | 6 | 1195 | |||
AK295315.1 | - | 5 | 641 | |||
AK299545.1 | - | 2 | 327 | |||
AL831915.1 | - | 5 | 767 | |||
BC025994.1 | - | 5 | 583 | 4822306 | ||
DQ063579.2 | - | 5 | 684 | |||
DQ975204.1 | - | 5 | 511 | |||
DQ996398.1 | - | 5 | 745 | |||
DQ996537.1 | - | 5 | 511 |