Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MECOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004991.3 | NP_004982.2 | 2 | 682 | ||
XM_005247213.3 | XP_005247270.1 | 2 | 688 | |||
XM_005247214.3 | XP_005247271.1 | 2 | 688 | |||
XM_005247215.3 | XP_005247272.1 | 2 | 687 | |||
XM_005247224.3 | XP_005247281.1 | 2 | 688 | |||
XM_005247225.3 | XP_005247282.1 | 2 | 687 | |||
XM_005247226.3 | XP_005247283.1 | 2 | 565 | |||
XM_017005877.1 | XP_016861366.1 | 2 | 687 | |||
GenBank mRNA | AK304098.1 | - | 2 | 628 | ||
AK308404.1 | - | 2 | 327 | |||
BC069498.1 | - | 2 | 531 | 7262213 | ||
CR541866.1 | - | 2 | 279 | |||
CR541886.1 | - | 2 | 279 | |||
U43292.1 | - | 2 | 586 | |||
U43293.1 | - | 1 | 242 |