Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 6 | 851 | ||
NM_005426.2 | NP_005417.1 | 7 | 983 | |||
XM_011544268.1 | XP_011542570.1 | 5 | 712 | |||
XM_011544269.2 | XP_011542571.1 | 3 | 370 | |||
XM_017002220.1 | XP_016857709.1 | 7 | 992 | |||
GenBank mRNA | AJ318888.2 | - | 6 | 796 | ||
AK294432.1 | - | 6 | 703 | |||
BC040247.1 | - | 4 | 327 | 4454266 | ||
BC046150.1 | - | 6 | 794 | |||
BC058918.1 | - | 6 | 809 | 4537206 | ||
U58334.1 | - | 7 | 928 |