Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005732.3 | NP_005723.2 | 4 | 787 | ||
GenBank mRNA | AF057299.1 | - | 4 | 707 | ||
AF057300.1 | - | 4 | 642 | |||
AK290597.1 | - | 4 | 729 | |||
AK307439.1 | - | 4 | 462 | |||
BC062603.1 | - | 4 | 704 | |||
BC073850.1 | - | 4 | 704 | |||
BC108282.1 | - | 4 | 724 | |||
BC136436.1 | - | 4 | 679 | 9020425 | ||
DQ655928.2 | - | 4 | 390 | |||
U63139.1 | - | 4 | 774 |