Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286123.1 | NP_001273052.1 | 11 | 1909 | ||
NM_001286125.1 | NP_001273054.1 | 10 | 1796 | |||
NM_005835.3 | NP_005826.1 | 11 | 2211 | |||
XM_005248784.2 | XP_005248841.1 | 12 | 2359 | |||
XM_006714949.3 | XP_006715012.1 | 12 | 2096 | |||
XM_006714950.2 | XP_006715013.1 | 11 | 1986 | |||
XM_006714951.1 | XP_006715014.1 | 11 | 2246 | |||
XM_017010159.1 | XP_016865648.1 | 11 | 2249 | |||
GenBank mRNA | U90544.1 | - | 11 | 2096 |