Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005956.3 | NP_005947.3 | 11 | 1451 | ||
GenBank mRNA | AK302747.1 | - | 11 | 1461 | ||
AK309266.1 | - | 10 | 1064 | |||
AK312361.1 | - | 11 | 1156 | |||
BC009806.2 | - | 11 | 1117 | 3508998 | ||
BC050420.1 | - | 11 | 1142 | 5788176 | ||
J04031.1 | - | 11 | 1117 |