Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289032.1 | NP_001275961.1 | 4, 5 | 1489 | ||
NM_001289033.1 | NP_001275962.1 | 4, 5 | 1334 | |||
NM_006215.3 | NP_006206.2 | 4, 5 | 1174 | |||
GenBank mRNA | AK309586.1 | - | 4, 5 | 1467 | ||
AK313196.1 | - | 4, 5 | 1152 | |||
BC014992.1 | - | 4, 5 | 1148 | 4866432 | ||
BX248009.1 | - | 4, 5 | 1249 | |||
BX248760.1 | - | 4, 5 | 1137 | |||
L19684.1 | - | 3, 4 | 1082 |