Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006345.3 | NP_006336.3 | 2 | 382 | ||
XM_017007654.1 | XP_016863143.1 | 2 | 416 | |||
GenBank mRNA | AF006621.2 | - | 2 | 356 | ||
AK225470.1 | - | 2 | 319 | |||
AK299917.1 | - | 2 | 358 | |||
AK312838.1 | - | 2 | 382 | |||
AY319413.1 | - | 3 | 261 | |||
AY594282.1 | - | 2 | 236 | |||
BC000240.1 | - | 2 | 343 | |||
BC007732.2 | - | 2 | 343 | 4109702 | ||
BC016949.1 | - | 2 | 361 | 3850521 | ||
BG323207.1 | - | 382 | ||||
KU178526.1 | - | 2 | 236 | |||
KU178527.1 | - | 2 | 236 |