Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321703.1 | NP_001308632.1 | 6 | 740 | ||
NM_001321707.1 | NP_001308636.1 | 6 | 586 | |||
NM_001321711.1 | NP_001308640.1 | 6 | 668 | |||
NM_001321712.1 | NP_001308641.1 | 6 | 585 | |||
NM_001321713.1 | NP_001308642.1 | 5 | 481 | |||
NM_006540.3 | NP_006531.1 | 6 | 668 | |||
XM_017012961.1 | XP_016868450.1 | 6 | 655 | |||
XM_017012962.1 | XP_016868451.1 | 6 | 679 | |||
XM_017012963.1 | XP_016868452.1 | 6 | 1892 | |||
XM_017012964.1 | XP_016868453.1 | 6 | 947 | |||
XM_017012965.1 | XP_016868454.1 | 6 | 612 | |||
XM_017012966.1 | XP_016868455.1 | 7 | 876 | |||
XM_017012967.1 | XP_016868456.1 | 5 | 590 | |||
XM_017012968.1 | XP_016868457.1 | 7 | 677 | |||
XM_017012969.1 | XP_016868458.1 | 5 | 708 | |||
XM_017012970.1 | XP_016868459.1 | 5 | 7127 | |||
XM_017012971.1 | XP_016868460.1 | 5 | 839 | |||
XM_017012972.1 | XP_016868461.1 | 2 | 170 | |||
XM_017012973.1 | XP_016868462.1 | 6 | 655 | |||
GenBank mRNA | AK308426.1 | - | 6 | 633 | ||
BC114383.1 | - | 5 | 535 | 40082387 | ||
X97674.1 | - | 6 | 613 |