Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037535.2 | NP_001032624.1 | 6 | 1478 | ||
NM_001037536.2 | NP_001032625.1 | 6 | 1475 | |||
NM_001037540.2 | NP_001032629.1 | 8 | 1708 | |||
NM_006746.5 | NP_006737.2 | 7 | 1627 | |||
XM_005274578.2 | XP_005274635.1 | 8 | 1703 | |||
XM_005274579.2 | XP_005274636.1 | 7 | 1616 | |||
XM_006724508.3 | XP_006724571.1 | 7 | 4608 | |||
XM_006724509.3 | XP_006724572.1 | 6 | 4524 | |||
XM_011545564.2 | XP_011543866.1 | 7 | 4611 | |||
XM_017029723.1 | XP_016885212.1 | 6 | 4527 | |||
GenBank mRNA | AF160728.1 | - | 6 | 1415 | ||
BC026159.2 | - | 7 | 1606 | 4827364 | ||
BC105028.1 | - | 4 | 1083 | 8144031 | ||
BX647757.1 | - | 5 | 3393 |