Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130060.1 | NP_001123532.1 | 12 | 2137 | ||
NM_006856.2 | NP_006847.1 | 12 | 2200 | |||
XM_005268587.3 | XP_005268644.1 | 12 | 2152 | |||
XM_017018722.1 | XP_016874211.1 | 12 | 3340 | |||
GenBank mRNA | AK302907.1 | - | 8 | 1215 | ||
BC136302.1 | - | 12 | 2086 | 9020289 | ||
X52943.1 | - | 12 | 2145 |