Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006885.3 | NP_008816.3 | 2 | 2381 | ||
XM_005255957.3 | XP_005256014.1 | 2 | 10859 | |||
GenBank mRNA | L32832.1 | - | 2 | 2381 |