Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002294.2 | NP_001002294.1 | 7 | 1107 | ||
NM_001319173.1 | NP_001306102.1 | 8 | 1217 | |||
NM_001319174.1 | NP_001306103.1 | 6 | 918 | |||
NM_006894.5 | NP_008825.4 | 7 | 1090 | |||
XM_011509345.2 | XP_011507647.1 | 8 | 1238 | |||
GenBank mRNA | AK223166.1 | - | 7 | 1099 | ||
AK296105.1 | - | 6 | 892 | |||
AK298406.1 | - | 8 | 1202 | |||
AK313197.1 | - | 7 | 1080 | |||
BC032016.1 | - | 7 | 1042 | 5175615 | ||
KU178077.1 | - | 6 | 996 | |||
M83772.1 | - | 7 | 1132 | |||
Z47552.1 | - | 7 | 1089 |