Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007053.3 | NP_008984.1 | 5 | 781 | ||
NR_103845.1 | 3 | 309 | ||||
XM_005272929.3 | XP_005272986.1 | 4 | 809 | |||
XM_011509104.2 | XP_011507406.1 | 5 | 915 | |||
GenBank mRNA | AF060981.1 | - | 5 | 704 | ||
AK128370.1 | - | 3 | 320 | |||
BC014465.1 | - | 5 | 707 | 4849145 | ||
CD638675.1 | - | 3 | 312 | |||
CR541867.1 | - | 3 | 489 | |||
CR541888.1 | - | 3 | 489 | |||
EU016100.1 | - | 3 | 489 | |||
EU016101.1 | - | 2 | 162 |