Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308197.1 | NP_001295126.1 | 5 | 1301 | ||
NM_001308204.1 | NP_001295133.1 | 5 | 1127 | |||
NM_001308205.1 | NP_001295134.1 | 5 | 1151 | |||
NM_007107.4 | NP_009038.1 | 5 | 1262 | |||
GenBank mRNA | AF110647.1 | - | 5 | 1206 | ||
BC017203.1 | - | 5 | 1207 | 3618589 |