Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308197.1 | NP_001295126.1 | 5 | 2977 | ||
NM_001308204.1 | NP_001295133.1 | 5 | 2803 | |||
NM_001308205.1 | NP_001295134.1 | 5 | 2827 | |||
NM_007107.4 | NP_009038.1 | 5 | 2938 | |||
GenBank mRNA | AF110647.1 | - | 5 | 2878 | ||
CX786292.1 | - | 1 | 338 |