Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 9 | 1616 | ||
NM_012470.3 | NP_036602.1 | 9 | 1616 | |||
NR_034053.2 | 10 | 1777 | ||||
XM_011515989.2 | XP_011514291.1 | 9 | 1218 | |||
GenBank mRNA | AF145029.1 | - | 9 | 1326 | ||
AJ133769.1 | - | 9 | 1213 | |||
AK001263.1 | - | 2 | 170 | |||
AK225999.1 | - | 9 | 1587 | |||
AK312580.1 | - | 9 | 1343 | |||
BC068530.1 | - | 9 | 1344 |