Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013351.1 | NP_037483.1 | 6 | 1874 | ||
XM_011524698.2 | XP_011523000.1 | 7 | 1941 | |||
GenBank mRNA | AF093098.1 | - | 6 | 1874 | ||
AK223634.1 | - | 6 | 1867 | |||
BC039739.2 | - | 6 | 1839 | 6067371 |