Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013386.4 | NP_037518.3 | 10 | 2714 | ||
NM_213651.2 | NP_998816.1 | 10 | 2625 | |||
XM_017001104.1 | XP_016856593.1 | 6 | 1970 | |||
GenBank mRNA | AF123303.1 | - | 10 | 2471 | ||
AJ619961.1 | - | 10 | 2558 | |||
AJ619987.1 | - | 10 | 2503 | |||
AL050209.1 | - | 1 | 512 | |||
BC014519.1 | - | 10 | 2570 | 3842039 | ||
BC029377.1 | - | 1 | 498 | |||
BC068010.1 | - | 1 | 653 | |||
BC068561.1 | - | 10 | 2603 | 5259336 |