Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076683.1 | NP_001070151.1 | 8 | 977 | ||
NM_001076684.2 | NP_001070152.1 | 8 | 1063 | |||
NM_014233.3 | NP_055048.1 | 9 | 1174 | |||
NR_045058.1 | 9 | 1371 | ||||
XM_006722059.3 | XP_006722122.1 | 9 | 1213 | |||
XM_006722060.2 | XP_006722123.1 | 8 | 957 | |||
XM_006722061.2 | XP_006722124.1 | 9 | 1041 | |||
XM_017024995.1 | XP_016880484.1 | 9 | 1222 | |||
XM_017024996.1 | XP_016880485.1 | 9 | 1174 | |||
XM_017024997.1 | XP_016880486.1 | 9 | 1366 | |||
XM_017024998.1 | XP_016880487.1 | 9 | 1082 | |||
XM_017024999.1 | XP_016880488.1 | 8 | 957 | |||
XM_017025000.1 | XP_016880489.1 | 8 | 1015 | |||
XM_017025001.1 | XP_016880490.1 | 8 | 1063 | |||
XM_017025002.1 | XP_016880491.1 | 8 | 943 | |||
XM_017025003.1 | XP_016880492.1 | 8 | 953 | |||
XM_017025004.1 | XP_016880493.1 | 8 | 945 | |||
GenBank mRNA | AK291733.1 | - | 9 | 999 | ||
AK292518.1 | - | 9 | 1371 | |||
AK292577.1 | - | 9 | 1259 | |||
AK296917.1 | - | 3 | 287 | |||
AK298008.1 | - | 6 | 589 | |||
AK308321.1 | - | 7 | 1040 | |||
BC031423.1 | - | 7 | 1001 | |||
BC042297.1 | - | 8 | 968 | 4509695 | ||
X53390.1 | - | 8 | 1025 | |||
X53461.1 | - | 8 | 1025 | |||
X56687.1 | - | 8 | 946 |