Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014252.3 | NP_055067.1 | 7 | 1293 | ||
GenBank mRNA | AF112968.1 | - | 7 | 1130 | ||
AF177333.1 | - | 7 | 1095 | |||
AK296862.1 | - | 8 | 945 | |||
BC002702.2 | - | 7 | 1207 | 3614008 |