Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014423.3 | NP_055238.1 | 21 | 4009 | ||
XM_005271963.4 | XP_005272020.1 | 22 | 4073 | |||
XM_005271964.4 | XP_005272021.1 | 15 | 2721 | |||
XM_006714587.3 | XP_006714650.1 | 20 | 3922 | |||
GenBank mRNA | AF197927.1 | - | 21 | 3938 | ||
AF213987.2 | - | 21 | 3666 | |||
BC137226.1 | - | 21 | 3907 | 9021223 | ||
BC144538.1 | - | 22 | 3924 |