Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308190.1 | NP_001295119.1 | 2, 3 | 285 | ||
NM_002099.7 | NP_002090.4 | 3, 4 | 384 | |||
XM_017008134.1 | XP_016863623.1 | 3, 4 | 388 | |||
XM_017008135.1 | XP_016863624.1 | 4, 5 | 451 | |||
XM_017008136.1 | XP_016863625.1 | 2, 3 | 353 | |||
GenBank mRNA | AK290561.1 | - | 3, 4 | 349 | ||
BC005319.1 | - | 3, 4 | 290 | 3930700 | ||
BC013328.1 | - | 3, 4 | 271 | 4134368 | ||
DA537533.1 | - | 3, 4 | 349 | |||
DA539682.1 | - | 3, 4 | 385 | |||
EU338231.1 | - | 4, 5 | 353 | |||
EU338232.1 | - | 3, 4 | 270 | |||
EU338233.1 | - | 4, 5 | 353 | |||
EU338234.1 | - | 2, 3 | 177 | |||
GU347002.1 | - | 4, 5 | 377 | |||
GU347003.1 | - | 4, 5 | 456 | |||
GU347004.1 | - | 4, 5 | 434 | |||
GU347006.1 | - | 3, 4 | 276 | |||
GU347008.1 | - | 2, 3 | 171 | |||
L31860.1 | - | 3, 4 | 288 | |||
M12857.1 | - | 3, 4 | 243 | |||
M36281.1 | - | 3, 4 | 243 | |||
X08054.1 | - | 3, 4 | 289 | |||
X51798.1 | - | 3, 4 | 257 |