Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105538.1 | NP_001099008.1 | 7 | 901 | ||
NM_014520.3 | NP_055335.2 | 7 | 901 | |||
XM_011523616.2 | XP_011521918.1 | 7 | 893 | |||
GenBank mRNA | AF147709.1 | - | 7 | 864 | ||
AK303150.1 | - | 7 | 869 | |||
AK307948.1 | - | 7 | 900 | |||
AL136595.1 | - | 6 | 609 | |||
BC050546.1 | - | 7 | 844 | 5495483 |