Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014571.3 | NP_055386.1 | 5 | 1493 | ||
XM_005270745.2 | XP_005270802.1 | 5 | 1547 | |||
GenBank mRNA | BC006087.1 | - | 5 | 1463 | 2964729 |