Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271711.1 | NP_001258640.1 | 9 | 1601 | ||
NM_001271712.1 | NP_001258641.1 | 8 | 1377 | |||
NM_014580.4 | NP_055395.2 | 10 | 1747 | |||
NR_073416.1 | 7 | 1180 | ||||
XM_006717084.3 | XP_006717147.1 | 10 | 1418 | |||
XM_011518602.2 | XP_011516904.1 | 10 | 1663 | |||
XM_011518603.2 | XP_011516905.1 | 8 | 1402 | |||
XM_011518604.2 | XP_011516906.1 | 8 | 1326 | |||
XM_017014653.1 | XP_016870142.1 | 7 | 1221 | |||
GenBank mRNA | AB209112.1 | - | 4971 | |||
AF289587.1 | - | 9 | 1509 | |||
AJ245937.1 | - | 10 | 1657 | |||
BC019043.2 | - | 10 | 1699 | 4641145 |