Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015103.2 | NP_055918.2 | 33, 34 | 5553 | ||
XM_011512588.1 | XP_011510890.1 | 33, 34 | 5468 | |||
XM_011512589.1 | XP_011510891.1 | 30, 31 | 5078 | |||
XM_011512592.1 | XP_011510894.1 | 21, 22 | 2636 | |||
GenBank mRNA | AB014520.2 | - | 33, 34 | 5632 | ||
AK026388.1 | - | 9, 10 | 1050 | |||
AK299425.1 | - | 12, 13 | 1388 | |||
AY116661.1 | - | 33, 34 | 5633 | |||
BC003526.1 | - | 11, 12 | 1300 | |||
BC011848.2 | - | 10, 11 | 1243 | |||
BC044228.1 | - | 1, 2 | 278 | |||
BC150280.1 | - | 33, 34 | 5632 | 9007263 |