Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015305.3 | NP_056120.2 | 7 | 1718 | ||
XM_017021115.1 | XP_016876604.1 | 8 | 1766 | |||
GenBank mRNA | AB018302.1 | - | 7 | 1615 | ||
AK301589.1 | - | 3 | 285 | |||
AL137268.1 | - | 7 | 1616 | |||
BC028714.2 | - | 7 | 1665 | 4830897 |