Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016245.4 | NP_057329.3 | 2 | 499 | ||
XM_011532021.1 | XP_011530323.1 | 2 | 448 | |||
GenBank mRNA | AF126780.1 | - | 2 | 454 | ||
AF273056.1 | - | 2 | 352 | |||
AK075348.1 | - | 2 | 383 | |||
AY358553.1 | - | 2 | 366 | |||
BC008650.1 | - | 2 | 389 | 3854750 | ||
BC014327.1 | - | 2 | 365 | 3961721 | ||
BC016367.1 | - | 2 | 396 | 4133318 | ||
BC021673.1 | - | 2 | 370 | 3827935 | ||
BC036001.1 | - | 2 | 390 | 4715042 | ||
DA265697.1 | - | 2 | 498 |