Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016322.3 | NP_057406.2 | 8 | 3025 | ||
GenBank mRNA | AF052113.1 | - | 1 | 1471 | ||
AK000447.1 | - | 1 | 1339 | |||
AK023524.1 | - | 8 | 2807 | |||
AK090889.1 | - | 8 | 2824 | |||
AL110256.1 | - | 1 | 1408 | |||
AL162081.1 | - | 8 | 2751 |