Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016322.3 | NP_057406.2 | 8 | 3844 | ||
GenBank mRNA | AL162081.1 | - | 8 | 3570 |