Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278603.1 | NP_001265532.1 | 7 | 998 | ||
NM_017426.3 | NP_059122.2 | 8 | 1142 | |||
NR_103780.1 | 7 | 915 | ||||
NR_103781.1 | 7 | 915 | ||||
NR_103782.1 | 8 | 912 | ||||
XM_006714237.1 | XP_006714300.1 | 9 | 1119 | |||
XM_011532033.2 | XP_011530335.1 | 8 | 1085 | |||
XM_011532034.2 | XP_011530336.1 | 7 | 1385 | |||
XM_017008294.1 | XP_016863783.1 | 8 | 1529 | |||
XM_017008295.1 | XP_016863784.1 | 7 | 1302 | |||
GenBank mRNA | AK001517.1 | - | 8 | 1024 | ||
AK223352.1 | - | 8 | 1025 | |||
AK299442.1 | - | 7 | 797 | |||
AK300036.1 | - | 7 | 880 | |||
AK307571.1 | - | 8 | 802 | |||
AK315160.1 | - | 8 | 1022 | |||
AL137650.1 | - | 1 | 141 | |||
BC012559.1 | - | 8 | 1039 | |||
BX538002.1 | - | 7 | 815 |