Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017429.2 | NP_059125.2 | 9 | 1499 | ||
XM_017023287.1 | XP_016878776.1 | 9 | 1548 | |||
XM_017023289.1 | XP_016878778.1 | 5 | 613 | |||
GenBank mRNA | AF294900.1 | - | 9 | 1499 | ||
AK001592.1 | - | 9 | 1499 | |||
AK296011.1 | - | 8 | 1371 | |||
BC126210.1 | - | 9 | 1401 | 8991926 | ||
BC126212.1 | - | 9 | 1401 | 8991928 |