Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 1337 | ||
XM_017001691.1 | XP_016857180.1 | 3 | 1242 | |||
GenBank mRNA | AF374386.1 | - | 3 | 1256 | ||
AK000319.1 | - | 2 | 1058 | |||
AK024774.1 | - | 1 | 757 | |||
AK311305.1 | - | 4 | 1122 | |||
BC001959.1 | - | 3 | 1304 | 3536977 | ||
BC078177.1 | - | 3 | 1284 | 6526331 | ||
BC085018.1 | - | 3 | 1307 | 6650445 |